Optical adhesive PCR/qPCR film for Piko® UTW plates
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ASF-0020 ASF-0020/100
731-0404EA 2470 DKK
731-0404 ADVAASF-0020/100
Optical adhesive PCR/qPCR film for Piko® UTW plates
Forsegling Mikrotiterpladetætninger
Clear, 24-well format Piko® PCR/qPCR film seal allows high transmission of fluorescence across the range of commonly used fluorophores. When used with a Piko® thermal cycler or PikoReal® Real-Time PCR system, sample volumes as low as 5 µl can be used with minimal evaporative loss.
  • Optical clarity with minimal auto-fluorescence for Real-Time PCR applications
  • Tight sealing for sample volumes as low as 5 µl
  • Adhesive backing allows easy application without costly sealing devices
  • Cryo-compatible adhesive is effective to −80 °C