PCR plates, 96-well, full skirted, barcoded
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BC-0800/W BC-0800
ADVABC-0800/WEA 1560 DKK
ADVABC-0800/W ADVABC-0800
PCR plates, 96-well, full skirted, barcoded
Mikroplader PCR plader
The full-skirted design of these PP plates is robotic-friendly, allowing for automated processes.
  • ANSI footprint and stackable for use in automated systems
  • Low profile to reduce dead space and increase PCR efficiency
  • Cut corner: H1
  • Raised rim design around each well enables secure sealing and safeguards against evaporation
  • Uniform, thin well walls deliver maximum and consistent heat transfer
The barcodes are the industry-standard code 128. The label itself can withstand temperatures from -196 up to 120 °C to ensure that the barcodes stay on the plate throughout your entire workflow. It also has a special coating to virtually eliminate any "rubbing off" of the code during handling. The dual barcodes are located on the opposite numeric and opposite alpha sides of the plate.