Transport tubes, Axygen®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SCT-10ML SCT-5ML SCT-5ML-S ST-10ML ST-5ML
AXYGSCT-10MLEA 1770 DKK
AXYGSCT-10ML AXYGSCT-5ML AXYGSCT-5ML-S 212-0346 212-0357
Transport tubes, Axygen®
Rør Reagensglas
PP, translucent.
  • Graduation marks at 1,0 ml intervals
  • Large, frosted writing area for sample indentification
  • Secure fit between tube and cap provides a leak resistant seal
Universal tubes for a wide range of applications.
Bestil nu
Specifikationer
Environmentally Preferable Lav fremstillingspåvirkning
Miljøcertificering I-RECs, ISO 14001 - Reynosa