Gel permeation chromatography columns, PL Rapide
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PL1113-3500 PL1120-3830 PL1149-3800 PL1113-3100 PL1113-3120 PL1113-3300 PL1013-2100 PL1013-2120 PL1013-2300 PL1013-2500 PL1020-2830 PL1049-2800
VARIPL1113-3500EA 22610 DKK
VARIPL1113-3500 VARIPL1120-3830 VARIPL1149-3800 VARIPL1113-3100 VARIPL1113-3120 VARIPL1113-3300 VARIPL1013-2100 VARIPL1013-2120 VARIPL1013-2300 VARIPL1013-2500 VARIPL1020-2830 VARIPL1049-2800
Gel permeation chromatography columns, PL Rapide
Kromatografikolonner
These columns are ideal for high speed applications such as high throughput screening, process monitoring or tracking changes in MW distributions where time is the most critical factor in the analysis.
  • Analysis in less than ten minutes that saves time
  • Significantly increased sample throughput improves efficiency
  • Reduced solvent consumption and disposal costs save money
  • Available in L, M and H versions for low, medium and high molecular weights and also F version for flow injection analysis
  • Columns cover the complete spectrum of molecular weights and are available for the analysis of both organic and water soluble polymers
Rapid GPC is an excellent tool for screening polymer MWD for trend analysis. Short PL Rapide columns reduce analysis times while maintaining the excellent solvent compatibility and mechanical stability.