Capillary columns, VF-5 Pesticides and VF-1701 Pesticides
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CP9070 CP9071 CP9072 CP9073 CP9074 CP9075
VARICP9070EA 12920 DKK
VARICP9070 VARICP9071 VARICP9072 VARICP9073 VARICP9074 VARICP9075
Capillary columns, VF-5 Pesticides and VF-1701 Pesticides
Kromatografikolonner
These columns are specially designed for the determination of trace levels of pesticide residue.
  • Tested with key pesticides for improved efficiency
  • Highly inert for enhanced detection
  • Proven performance with ECD and MS detection for maximum productivity
VF-Pesticides columns benefit from ultra low bleed FactorFour technology to improve sensitivity. VF-1701 Pesticides deliver up to 8 times lower bleed than other columns used for pesticide analysis.