Multi-channel pipettes, variable volume, Acura® manual 855
Leverandør: Socorex Isba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
855.12.350 855.12.050 855.12.010 855.08.350 855.08.050 855.08.010 855.08.200 855.12.200 855.08.100
613-0572EA 5880 DKK
613-0572 613-0570 613-0569 613-0568 613-0566 613-0565 613-0567 613-0571 SOCO855.08.100
Multi-channel pipettes, variable volume, Acura® manual 855
Pipetter Automatiske pipetter
These pipettes are equipped with a precision setting mechanism and therefore continuously adjustable down to a very fine volume resolution. The line offers 8- and 12-channel models with volumes ranging from 0,5 to 350 µl.
  • Single-handed volume setting
  • Click stops, together with freely rotating Smartie plunger button cap secure setting stability while pipetting
  • Extend pipetting possibilities in microplates
  • Completely autoclavable