Tubing, PVC
Leverandør: MURRAY S
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
L800/01 L800/02 L800/03 L800/04 L800/05 L800/06 L800/07
MURRL800/01EA 324 DKK
MURRL800/01 MURRL800/02 MURRL800/03 MURRL800/04 MURRL800/05 MURRL800/06 MURRL800/07
Tubing, PVC
Slanger Overførselsslange
PVC, clear.
  • Very elastic grade
  • Non toxic
  • Can be sterilised up to 121 °C
This tubing is very flexible and will retain its bore on bending.