PIG® poly spill containment pallets
Leverandør: NEW PIG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PAK210-WOD PAK605-BK-WOD
121-0188EA 4480 DKK
121-0188 121-0183
PIG® poly spill containment pallets
Spild materialer
These economical pallets are strong enough to handle long-term storage of fully loaded steel drums.
  • Low-density polyethylene (LDPE) construction resists UV rays, rust, corrosion and most chemicals
  • Molded-in sump catches leaks, drips and spills to help you comply with regulations and keep your storage areas clean and safe
  • Strong enough to support four fully loaded steel or poly drums
  • Textured grating adds traction and lifts out for easy sump access
  • 2-way forklift entry allows easy movement from either direction (unloaded only)
Nestable design permits stacking when not in use; lowers cost of bulk shipping.