HPLC columns, Syncronis™ aQ
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
97305-254630 97305-154630 97302-052130 97305-104630 97305-102130 97305-053030 97305-014001 97305-013001 97305-012101 97302-102130 97303-104630 97303-154630 97303-103030
HYPE97305-254630EA 9560 DKK
HYPE97305-254630 HYPE97305-154630 HYPE97302-052130 HYPE97305-104630 HYPE97305-102130 HYPE97305-053030 HYPE97305-014001 HYPE97305-013001 HYPE97305-012101 HYPE97302-102130 HYPE97303-104630 HYPE97303-154630 HYPE97303-103030
HPLC columns, Syncronis™ aQ
Kromatografikolonner
These polar endcapped C18 columns provide a controlled interaction mechanism by which polar analytes can be retained and resolved.
  • Stable in 100% aqueous mobile phase
  • Enhanced retention of polar compounds
Surface area: 320 m²/g
Carbon load: 19%
pH range: 2 - 8