BCR® Cleanroom Bond Paper
Leverandør: BERKSHIRE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B85A4WH250/10 B104A4W.250/10 B85A4PK.250/10 B104A3WH.250/5 B85A4GN.250/10 B104A5W.250/20 B85A4BL25010 B85A4YL25010 B85A3.BL.250/5 B68A4WH25010 B68A4BL25010 B104A4B25010 B85A3WH2505
BERKB85A4WH250/10EA 290 DKK
BERKB85A4WH250/10 BERKB104A4W.250/10 BERKB85A4PK.250/10 BERKB104A3WH.250/5 BERKB85A4GN.250/10 BERKB104A5W.250/20 BERKB85A4BL25010 818-0295 BERKB85A3.BL.250/5 BERKB68A4WH25010 BERKB68A4BL25010 BERKB104A4B25010 BERKB85A3WH2505
BCR® Cleanroom Bond Paper
Papir
BCR® Bond is made with the pure quality cellulose and coated to minimise particle and fibre release. They come in a variety of weights and colours to suit a particular need, and are recommended for use in ISO Class 3 to 7 cleanrooms.
  • Made of cellulose
  • Low in particles and fibres
  • Double-packed for cleanliness
  • Smooth surfaces reduce particle contamination