Watch glasses
Leverandør: MURRAY S
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
G850/01 G850/02 G850/03 G850/04 G850/05
MURRG850/01EA 463 DKK
MURRG850/01 MURRG850/02 MURRG850/03 MURRG850/04 MURRG850/05
Watch glasses
Urglas
Made of glass.
  • Good chemical and physical properties
  • Use in applications that do not involve high thermal stresses
  • Quick and easy to clean
With ground edges for covering beakers.