Shelf Kits with Racks and Boxes for Ultra-Low Temperature Freezers
Leverandør: REVCO TECHNOLOGIES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SK500 RSK600SD5 RSK500SD5 RSK400SD5 RSK300SD5 RSK700SD5 RSK13SD5 RSK700SD5 RSK600SD5 RSK500SD5 RSK400SD5 RSK300SD5 RSK23SD5 RSK17SD5 RSK600SD4 RSK300SD4 RSK400SD4 RSK500SD4
REVCSK500EA 2500 DKK
REVCSK500 REVCRSK600SD5 REVCRSK500SD5 REVCRSK400SD5 REVCRSK300SD5 REVCRSK700SD5 HERARSK13SD5 HERARSK700SD5 HERARSK600SD5 HERARSK500SD5 HERARSK400SD5 HERARSK300SD5 HERARSK23SD5 HERARSK17SD5 HERARSK600SD4 HERARSK300SD4 HERARSK400SD4 HERARSK500SD4
Shelf Kits with Racks and Boxes for Ultra-Low Temperature Freezers
Møbler Hylder
Maximize storage space with shelf kits with racks and boxes for Thermo Scientific™ ultra-low temperature freezers, featuring 50,8 mm boxes with dividers.
  • Stainless steel racks
  • Boxes
  • Cell dividers
  • Colour: White