Tip boxes, dispolab®
Leverandør: KARTELL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9756-06 9756
KART9756-06EA 664 DKK
KART9756-06 KART9756
Tip boxes, dispolab®
Pipette Tip Boxes and Racks
Empty boxes for use with refill tips.
  • Semitransparent boxes in PC have a horizontal sliding cover
  • Yellow boxes in PP feature a snap-on cover