Polyethylene glycol/oxide standards
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PL2070-1001 PL2070-1005 PL2070-1010 PL2070-2001 PL2070-2005 PL2070-2010 PL2070-3001 PL2070-3005 PL2070-3010 PL2070-4001 PL2070-4005 PL2070-4010 PL2070-5001 PL2070-5005 PL2070-5010 PL2070-6001 PL2070-6005 PL2070-6010 PL2070-7001 PL2070-7005 PL2070-7010 PL2070-8001 PL2070-8005 PL2070-8010 PL2070-9001 PL2070-9005 PL2070-9010 PL2071-0001 PL2071-0005 PL2071-0010 PL2071-1001 PL2071-1005 PL2083-6001 PL2083-6005 PL2083-7001 PL2083-8001 PL2083-9001 PL2084-0001 PL2071-1010 PL2084-1001 PL2084-2001 PL2084-2005 PL2084-2010 PL2071-2001 PL2071-3001 PL2071-3005 PL2083-1001 PL2083-1010 PL2083-2001 PL2083-2005 PL2083-2010 PL2083-3001 PL2083-3005 PL2083-3010 PL2083-4001 PL2083-4005 PL2083-5001 PL2083-5010
VARIPL2070-1001EA 2020 DKK
VARIPL2070-1001 VARIPL2070-1005 VARIPL2070-1010 VARIPL2070-2001 VARIPL2070-2005 VARIPL2070-2010 VARIPL2070-3001 VARIPL2070-3005 VARIPL2070-3010 VARIPL2070-4001 VARIPL2070-4005 VARIPL2070-4010 VARIPL2070-5001 VARIPL2070-5005 VARIPL2070-5010 VARIPL2070-6001 VARIPL2070-6005 VARIPL2070-6010 VARIPL2070-7001 VARIPL2070-7005 VARIPL2070-7010 VARIPL2070-8001 VARIPL2070-8005 VARIPL2070-8010 VARIPL2070-9001 VARIPL2070-9005 VARIPL2070-9010 VARIPL2071-0001 VARIPL2071-0005 VARIPL2071-0010 VARIPL2071-1001 VARIPL2071-1005 VARIPL2083-6001 VARIPL2083-6005 VARIPL2083-7001 VARIPL2083-8001 VARIPL2083-9001 VARIPL2084-0001 VARIPL2071-1010 VARIPL2084-1001 VARIPL2084-2001 VARIPL2084-2005 VARIPL2084-2010 VARIPL2071-2001 VARIPL2071-3001 VARIPL2071-3005 VARIPL2083-1001 VARIPL2083-1010 VARIPL2083-2001 VARIPL2083-2005 VARIPL2083-2010 VARIPL2083-3001 VARIPL2083-3005 VARIPL2083-3010 VARIPL2083-4001 VARIPL2083-4005 VARIPL2083-5001 VARIPL2083-5010
Polyethylene glycol/oxide standards
Standarder Kromatografi Standarder
These hydrophilic polymers are suitable for both aqueous SEC and organic GPC using the majority of polar organic solvents. The two types are chemically similar that can be used together across a wider molecular weight range with aqueous and organic polymers.
  • Simple to use kit that combines glycols and oxides to extend the MW range and cover more applications
  • Polyethylene oxide covers the high molecular weights and polyethylene glycol covers the lower molecular weight range
  • MW's selected to provide equidistant calibration points for greater accuracy