Vacuette® Blood Collection Sets
Leverandør: Greiner Bio-One
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
450092_50 450095_1000 450085_240 450081_1000 450082_50 450086_240 450085_24 450085V1_420
GREI450092_50EA 198 DKK
GREI450092_50 GREI450095_1000 720-2783 GREI450081_1000 GREI450082_50 GREI450086_240 GREI450085_24 GREI450085V1_420
Vacuette® Blood Collection Sets
Kliniske diagnose systemer Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
These safety blood collection sets are available in several versions – Either with or without Luer adapter, or with Luer-adapter and holder.
  • Safety mechanism activated whilst the tip is still in the vein
  • High level of protection
  • Acoustic signal 'click' indicates correct activation
Especially suitable for patients with difficult vein conditions or thin veins.