Jars, wide mouth, with screw cap
Leverandør: MURRAY S
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
P202/21 P202/22 P202/23 P202/24 P202/25 P202/26 P202/23G808/08 P202/26P515/35 P202/22G808/07 P202/24P515/35
MURRP202/21EA 909 DKK
MURRP202/21 MURRP202/22 MURRP202/23 MURRP202/24 MURRP202/25 MURRP202/26 MURRP202/23G808/08 MURRP202/26P515/35 MURRP202/22G808/07 MURRP202/24P515/35
Jars, wide mouth, with screw cap
Dåser Dåser, allround
Jars, made of HDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • Available complete with screw cap or without screw cap
Wide mouth universal containers for creams, semi solid substances, crystals, powders and granulates.