GC injection port septa
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
702602 702604 702605 702606 702609 702610 702611 702612 702613 702619 702620 702621 702622 702623 702625 702626 702627 702628 702632 702659 702646 702647 702648 702649 702656 702657 702658 702660 702661 702662 702663
MANA702602EA 783 DKK
MANA702602 MANA702604 MANA702605 MANA702606 MANA702609 MANA702610 MANA702611 MANA702612 MANA702613 MANA702619 MANA702620 MANA702621 MANA702622 MANA702623 MANA702625 MANA702626 MANA702627 MANA702628 MANA702632 MANA702659 MANA702646 MANA702647 MANA702648 MANA702649 MANA702656 MANA702657 MANA702658 MANA702660 MANA702661 MANA702662 MANA702663
GC injection port septa
Septa
GC septa with premium HT and standard quality.
Premium HT quality:
- Blister pack for cleanliness and easily handling
- CenterGuide™ for a centred needle insertion
- High inert and durable materials
- High temperature stability (300/400 °C)
Standard quality:
- Inert silicone materials
- Temperature stability from 200 to 320 °C
* Plug