Forceps
Leverandør: MURRAY S
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
E006/02 E006/01
232-0037EA 73.2 DKK
232-0037 232-0200
Forceps
Pincet
Stainless steel. Dissecting forceps with guide pin, which helps to keep the fine tips aligned.
  • Sharp, narrow flutes
  • Curved