Kontrolglas, ISOSLIDE®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.00408.0001 1.00425.0001 1.02560.0001 1.00380.0001
1.00408.0001EA 2350 DKK
1.00408.0001 1.00425.0001 1.02560.0001 1.00380.0001
Kontrolglas, ISOSLIDE®
Objektglas Microscopy Control Slides
ISOSLIDE® control slides are designed for use with new as well as existing products.
  • Shelf life
  • Prepared exclusively from animal material
  • All slides in a box are from the same batch and are prepared from one block
ISOSLIDE® control slides help optimise and standardise staining results by allowing typical control material to be compared with a laboratory’s own specimens. Typical material is prepared and sliced using standard procedures. One slide is already stained by the Merck staining procedure, while the other 24 are unstained paraffin sections.More Product Information