Splash heads, with sinter, Quickfit®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SH16/29 SH16/19 SH16/24
201-0576EA 1640 DKK
201-0576 BIBBSH16/19 BIBBSH16/24
Splash heads, with sinter, Quickfit®
Destillations komponenter Hoveder
PYREX® borosilicate glas
  • Prevents possible contamination of the condenser from 'bumping' solvents
  • Features a coarse porosity 1 sinter to reduce carry over
  • Splash heads feature a 29/32 socket
For use with rotary evaporators.