Splash heads, anti-climb, Quickfit®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SH15/29 SH15/24 SH15/19
201-0573EA 1500 DKK
201-0573 201-0578 BIBBSH15/19
Splash heads, anti-climb, Quickfit®
Destillations komponenter Hoveder
PYREX® borosilicate glass.
  • Protects against liquid ingress into the condenser
  • Useful for liquids that have tendency to foam
  • Splash heads feature a 29/32 socket
For use with rotary evaporators.