Micro cuvettes
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6610019500 6610021100
636-0170EA 8680 DKK
636-0170 VARI6610021100
Micro cuvettes
Kuvetter Spektrofotometerkuvetter
Black walled rectangular UV quartz cuvettes self masking to match the Cary 50 specifications.