PendoTech Single-Use Single-Channel Temperature Sensor Benchtop Transmitter, 4-20 mA Output, ABS Plastic Box; 24VDC
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR Item#MFLX19406-47
Your Organization's Price:
Optimize performance of your PendoTECH® Pressure Sensor.
  • Choose from a wide range of replacement parts and accessories
  • Produces a 4-20 mA signal that is linear with temperature
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
The single-channel temperature sensor benchtop transmitter easily connects to the PendoTECH temperature sensor. It features a rugged ABC plastic enclosure and LED front panel power indicator. Electrical connections are located on the back panel. The narrow 4-20mA output range from 0 to 70ºC ensures accurate and drift free linearization of the sensor output.
***Must order reusable temperature sensor cables (19406-14 for luer sensor and 19406-15 for hose barb sensors) separately.***
<b>Leveringsinformation: </b>Power cord and plug.
More Product Information
Other Products In this Family
See all Items in this Product Family