Micropump Drive for A-Mount pump heads, explosion-proof (XPRF), 3150 rpm, 115/230 VAC
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR Item#MFLX07144-95
Your Organization's Price:
A variety of low-cost drives for numerous applications.
  • Magnetic coupling minimizes friction—all horsepower is transmitted directly to the gears
  • Hundreds of hours of dependable leakproof service
These Economical Drives for Micropump A-Mount Pump Heads are designed with magnetic coupling to minimize friction. Choose from AC motor drives, DC motor drives, explosion-proof drives, or air-powered drives.<br /><br />Explosion-proof and air-powered models are ideal for safe pumping in hazardous areas. Explosion-proof drives are rated for NEMA Class 1, Groups C and D locations.
More Product Information
Other Products In this Family
See all Items in this Product Family