Masterflex® Transfer Tubing, Tygon® ND-100-65, 1/4" ID x 5/16" OD; 50 Ft
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR Item#MFLX95666-13
Your Organization's Price:
Drains completely.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Tubing size: 1/4" ID x 5/16" OD
<B>Formulation:</B> ND-100-65<br /><B>Applications:</B> Pharmaceutical and biotech research applications.<br /><B>Characteristics:</B>Non-DEHP, phthalate free. Non-wetted surface permits complete drainage. Bondable, weldable. Clear.<br /><B>Certification:</B> USP Class VI, REACH, ISO 10993<br /><B>Temperature range:</B> -44 to 165 °F (-42 to 74 °C)<br /><B>Sterilize:</B> Autoclave, ethylene oxide, or irradiation.
More Product Information
Other Products In this Family
See all Items in this Product Family