Masterflex® L/S® Gear Service Kits for 10 to 600 rpm Digital Modular Drives
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR Item#MFLX77300-01
Your Organization's Price:
Make the most of your Masterflex drives replacement parts and service kits.
  • Easy to install, these service kits avoid costly downtime of your gear pump head
  • Suitable for use with drives 77301-20 and -30.
Extend the life of your Masterflex pump with these replacement parts.
<b>Leveringsinformation: </b>Includes gear assembly (nylon gear/shaft), and gear case cover gasket.
More Product Information
Other Products In this Family
See all Items in this Product Family