Masterflex® Flow Display and Totalizer; Desktop Enclosure
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR Item#MFLX24507-25
Your Organization's Price:
Build a complete single-use flow sensor system customizable to your application.
  • Modular interchangeable flow sensor systems with single-use design
  • Quickly connect 5 to 30 V square wave pulse output for easy measurement readings
The Masterflex® single-use flow sensor system is comprised of four components. Select the flow sensor, flow tubes, barcode scanner, and output that suit your application needs for a customizable approach to single-use flow measurement.<br /><br />Four output options are available — digital to analog converter, metering pump control system, panel mount flow display with totalizer, and benchtop flow display with totalizer.<br /><br />Flow in your tubing is picked up via the IR beam in the sensor connection and converted into a 5 to 30 V square wave pulse output. <br /><br />The digital-to-analog converter converts the 5 to 30 V square wave pulse to a voltage, current, or frequency output. <br /><br />The metering pump control system allows for adjusting and controlling metering pumps, such as a peristaltic pump, in batch or flow processes. Control the channel of one pump with flow velocity and totalizing data read out. <br /><br />The panel and benchtop flow displays with totalizer features a large, backlit, 8-digit, LCD showing accurate flow readout.<br /><br />Flow tube sensor, flow tubes, and barcode scanner are sold separately.
More Product Information
Other Products In this Family
See all Items in this Product Family