PendoTech PDKTP-RS232 Single-Use RS232 Sensor Cable with 4-Wire Leads; 12 ft (3.7 m)
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR Item#MFLX19406-82
Your Organization's Price:
Optimize performance of your PendoTECH® Pressure Sensor.
  • Choose from a wide range of replacement parts and accessories
  • Produces a 4-20 milliamp signal that is linear with temperature
  • Connections to the transmitter are quick and convenient
  • Convenient mounting screws
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
Use this cable to connect to your PendoTECH PressureMAT™ data output to USB serial port.
***Must order reusable temperature sensor cables (19406-14 for luer sensor and 19406-15 for hose barb sensors) separately.***
<b>Leveringsinformation: </b>Power cord and plug.
More Product Information
Other Products In this Family
See all Items in this Product Family