PendoTech TT1-DR Single-Use DIN Rail Mounting Kit for Temperature Sensor Transmitter
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR Item#MFLX19406-46
Your Organization's Price:
Optimize performance of your PendoTECH® Pressure Sensor.
  • Choose from a wide range of replacement parts and accessories
  • Produces a 4-20 milliamp signal that is linear with temperature
  • Connections to the transmitter are quick and convenient
  • Convenient mounting screws
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
Connects flow through tubing to luer fitting single use pressure sensors.
***Must order reusable temperature sensor cables (19406-14 for luer sensor and 19406-15 for hose barb sensors) separately.***
<b>Leveringsinformation: </b>Power cord and plug.
More Product Information
Other Products In this Family
See all Items in this Product Family