Masterflex® L/S® Gear Service Kits for 1 to 100 rpm Drives
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR Item#MFLX07553-08
Your Organization's Price:
Make the most of your Masterflex drives replacement parts and service kits.
  • Easy to install, these service kits avoid costly downtime of your gear pump head
  • Suitable for use with Masterflex® L/S® 1 to 100 rpm drives
Extend the life of your Masterflex pump with these replacement parts.
<b>Leveringsinformation: </b>Includes two gear assemblies (large and small), two gear case cover gaskets, and one vial of high temperature silicone grease.
More Product Information
Other Products In this Family
See all Items in this Product Family