Laboratory bottle, glass, amber, protect, plastic safety coated, narrow neck, GL thread, UV protected, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
218065435
SCOT218065435EA 8090 DKK
SCOT218065435
Laboratory bottle, glass, amber, protect, plastic safety coated, narrow neck, GL thread, UV protected, DURAN®
Flasker Glasflasker
DURAN®, borosilicate glass 3.3, protect, amber, without screw cap and pouring ring.
  • Very good chemical resistance, high temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Purely external application of the colour ensures unchanged DURAN® properties on the interior
  • Glass Type I according to USP, EP and JP
  • Amber glass meets USP 671 and Eur. Ph. 3.2.1. for protection against UV light
Round, with graduation and DIN thread. The coating provides scratch, leak and splinter protection and is ideallyMore Product Information