Portable photometers, pHotoFlex®, WTW
Leverandør: WTW
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
251100 251110 251200 251210 251105 251205
665-0168EA 23850 DKK
665-0168 665-0169 705-0562 665-0187 705-0765 WTWA251205
Portable photometers, pHotoFlex®, WTW
Spektrofotometer og fotometer UV/Vis-spektrofotometre
Portable photometers suitable for a wide range of photometry, turbidity and pH applications in the areas of environmental monitoring, water treatment, wine and beverage industries and process control.
  • User guidance via display for easy operation without handbook
  • Integrated pH measurement with automatic temperature compensation (except STD model)
  • Battery operation with four AA batteries for approximately 3000 measurements
A LabStation upgrades the instruments into a small laboratory instrument, comprises a rechargeable battery pack (RB Flex) and LSdataMore Product Information