Low temperature BOD incubator, WiseCube®WIR
Leverandør: witeg Labortechnik
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
DH.WIR03250
WITGDH.WIR03250EA 57190 DKK
WITGDH.WIR03250
Low temperature BOD incubator, WiseCube®WIR
Inkubatorer BIR inkubatorer
Low temperature BOD incubator with superior temperature uniformity by forced convection.
  • Intelligent control of refrigerator for preventing overload of compressor
  • RS232C interface for monitoring and controlling with PC
  • Stainless steel chamber and teflon-coated shelves, corrosion resistant
  • Over temperature and current protection, sensor error detection and leakage breaking
  • Back-light LCD display, alarm and timer available
  • Locking mode supported for experimental safety.
  • CFC-free (134A) refrigeration system
  • Digital fuzzy control system implementing superior