BOD bottles, with robotic stopper, WHEATON®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
227580
WHEA227580EA 2780 DKK
WHEA227580
BOD bottles, with robotic stopper, WHEATON®
Flasker BOD-flasker
Made of Type I borosilicate glass.
  • With glass Robotic stopper
  • Large capacity
We recommend this 2 Liter BOD Bottle for long-term BOD and ultimate BOD analysis.