Natriumchlorid 26% in aqueous solution

Leverandør: VWR Collection
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Saline, NaCl, Sea salt, Halite

VWRS6039.5000EA 0 DKK
VWRS6039.5000
Natriumchlorid 26% in aqueous solution
Sodium chloride
Formel: NaCl
Massefylde: 1…1,2 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003477
CAS-nummer: 7647-14-5
EINECS: 231-598-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR