VWR®, Water Sampling Bottles, Sterile
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
331-0256EA 1940 DKK
331-0256 331-0267 331-0275 331-0277 VWRI331-0256.1 VWRI331-0267.1 VWRI331-0268.1 VWRI331-0269.1 VWRI331-0270.1 VWRI331-0271.1 VWRI331-0272.1 VWRI331-0273.1 VWRI331-0274.1 VWRI331-0275.1 VWRI331-0276.1 VWRI331-0277.1
VWR®, Water Sampling Bottles, Sterile
Flasker Flasker til vandprøvetagning
PET, virgin, food grade with PP caps.
  • Different coloured caps for easy indentification of test
  • Tamper-evident strip to ensure sampling integrity
  • Full traceability, batch number and individual bottle number on each label
  • Fully recyclable
  • Cap diameter: 32 mm
Lightweight, durable bottles. Caps designed to shield neck from contamination. Choice of dosed and undosed bottles. Rectangular bottles for better storage with limited space.
Blue cap: For natural, non chlorinated water without sodium thiosulphate.
Red cap: For chlorinated water dosed with 20More Product Information