Acetone 99,8 VLSI, Electronic Grade

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Propanon

VWRC6683.2500EA 1300 DKK
VWRC6683.2500
Acetone 99,8 VLSI, Electronic Grade
Acetone
Formel: CH₃COCH₃
Molekylvægt: 58.08 g/mol
Kogepunkt: 56.2 °C (1013 hPa)
Smeltepunkt: –95.4 °C
Massefylde: 0.792 g/cm³ (20 °C)
Flammepunkt: <–20 °C (closed cup)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008765
CAS-nummer: 67-64-1
EINECS: 200-662-2
UN nummer: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067

Bestil nu

Specifikationer Testresultater

Assay (GC) ≥ 99.8 %
Identification (IR) Passes test
Colouration ≤ 10 APHA
Particle size (> 0.50 μm) (/ml) (*) ≤ 200
Resistivity (MOhm-cm) ≥ 5
Aldehyde (as HCHO) ≤ 10 ppm
Ethanol (GC) ≤ 100 ppm
Evaporation residue ≤ 3 ppm
Free acid (as CH₃COOH) ≤ 20 ppm
Free alkali (as NH₃) ≤ 2 ppm
Methanol (GC) ≤ 0.05 %
Substances reducing KMnO₄ (as O) ≤ 2.5 ppm
Water ≤ 0.2 %
Ag (Silver) ≤ 10 ppb
Al (Aluminium) ≤ 50 ppb
As (Arsenic) ≤ 10 ppb
Au (Gold) ≤ 20 ppb
B (Boron) ≤ 10 ppb
Ba (Barium) ≤ 20 ppb
Be (Beryllium) ≤ 10 ppb
Bi (Bismuth) ≤ 20 ppb
Ca (Calcium) ≤ 100 ppb
Cd (Cadmium) ≤ 10 ppb
Co (Cobalt) ≤ 10 ppb
Cr (Chromium) ≤ 10 ppb
Cu (Copper) ≤ 10 ppb
Fe (Iron) ≤ 10 ppb
Ga (Gallium) ≤ 10 ppb
In (Indium) ≤ 10 ppb
K (Potassium) ≤ 20 ppb
Li (Lithium) ≤ 10 ppb
Mg (Magnesium) ≤ 20 ppb
Mn (Manganese) ≤ 10 ppb
Mo (Molybdenum) ≤ 10 ppb
Na (Sodium) ≤ 100 ppb
Ni (Nickel) ≤ 10 ppb
Pb (Lead) ≤ 10 ppb
Pt (Platinum) ≤ 50 ppb
Sb (Antimony) ≤ 10 ppb
Sn (Tin) ≤ 20 ppb
Sr (Strontium) ≤ 10 ppb
Ti (Titanium) ≤ 20 ppb
Tl (Thallium) ≤ 10 ppb
V (Vanadium) ≤ 10 ppb
Zn (Zinc) ≤ 20 ppb
Zr (Zirconium) ≤ 20 ppb
Additional information (*): Particle size (> 0.50 μm) (/ml) in raw material before filling


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR