Raw fibre extractors
Leverandør: VELP SCIENTIFIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SA30520201 SA30520200 SA30530200 SA30530201
VELPSA30520201EA 48030 DKK
VELPSA30520201 VELPSA30520200 VELPSA30530200 VELPSA30530201
Raw fibre extractors
Ekstraktorer Systemer til fiberbestemmelse
Raw fibre determination is useful for nutritional, economic and legislative aspects in the food industry, where fibre content is often requirement for labelling of packaged foods. The FIWE units provide a semi automatic process to meet requirements of ADF, NDF, Weende and Van Soest methodologies.
  • Rapid analysis, reliable results and high reproducibility
  • Performs single or sequential extraction including boiling, rinsing and filtration
  • Sample removal with air pump
  • Reagent discharge with peristaltic pump
Leveringsinformation: FIWE units supplied withMore Product Information