Vortex mixers, ZX4 and TX4
Leverandør: VELP SCIENTIFIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F202A0280 F202A0270 SA202A0280
444-2002EA 1620 DKK
444-2002 444-2001 VELPSA202A0280
Vortex mixers, ZX4 and TX4
Rystere Vortex Mixere
The ZX4 and TX4 are digital vortex mixers with adjustable stirring speed and two operating modes, sensor or continuous, for outstanding repeatability. Their ergonomic and innovative design along with the unit’s epoxy coated zinc alloy base gives excellent stability on the bench, 4 non-slip feet prevent “walking” during use. The technopolymer structure ensures optimum chemical resistance and enhanced handling.
  • Unique IR sensor mode, an infrared system detects the presence of the test tube and the vortex mixer automatically starts vibrating. This reduces