Overhead omrørere, elektroniske, OHS digital og advance
Leverandør: VELP SCIENTIFIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F20100493 F20100494 F20100480 F20100481 F20100482 F20100490 F20100491 F20100492 SA20100492
441-0476EA 7830 DKK
441-0476 441-0475 441-0482 441-0481 441-0480 441-0479 441-0478 441-0477 VELPSA20100492
Overhead omrørere, elektroniske, OHS digital og advance
Omrørere Overhead omrørere
The OHS Series overhead stirrers are the ideal solution for the most demanding applications: The powerful brushless motor and microprocessor ensure excellent performance and make the VELP OHS Series overhead stirrers unlike any conventional stirrer.
  • Premium stirring power to complete any task
  • Brushless motor ensures maximum power up to 200 Ncm
  • OHS series maintain a constant torque at any speed (up to 2000 rpm)
  • Torque indicator to monitor viscosity changes
  • Integral timer
  • SpeedServo™ counter-reaction technology
  • Models with single- and double-speed gears