Capillary columns, VF-WAXms
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CP9206 CP9215 CP9216 CP9217 CP9218 CP9219 CP9220 CP9221 CP9222 CP9223 CP9225 CP9226 CP9228 CP9229 CP9201 CP9202 CP9203 CP9204 CP9205I5 CP9207 CP9210 CP9211 CP9212 CP9213 CP9214
VARICP9206EA 11650 DKK
VARICP9206 VARICP9215 VARICP9216 VARICP9217 VARICP9218 VARICP9219 VARICP9220 VARICP9221 VARICP9222 VARICP9223 VARICP9225 VARICP9226 VARICP9228 VARICP9229 VARICP9201 VARICP9202 VARICP9203 VARICP9204 VARICP9205I5 VARICP9207 VARICP9210 VARICP9211 VARICP9212 VARICP9213 VARICP9214
Capillary columns, VF-WAXms
Kromatografikolonner
VF-WAXms is a high performance column for applications in the food, flavours and fragrances markets, and especially where trace analyses are required.
  • Specially designed for MS for more accurate results with polar compounds
  • Operating temperature range of 20 to 250 °C for maximum flexibility
  • Better signal-to-noise ratio for trace analyses improves productivity
  • Provides increased sensitivity, extended column lifetime and greater accuracy, even at higher temperatures
  • Highly inert
VF-WAXms columns give better chromatograms, enhancing critical pair separation.