Disposable particle filtering respirators
Leverandør: DELTALYOVALMY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VSPX353-07C2
VALMVSPX353-07C2EA 0 DKK
VALMVSPX353-07C2
Disposable particle filtering respirators
Åndedrætsværn Engangsmasker Partikelmasker
These comfortable disposable FFP2/FFP3 masks are made from 100% recyclable material and protects against odors caused by organic gases and acids.
  • Airtightness is guaranteed thanks to its patented nose seal
  • Low breathing resistance
  • Less blur and perspiration
  • Latex-, silicone- and PVC-free
Certificeringer: EN 149:2001+A1:2009