VARIO® MD 12C NT VARIO select Chemistry Diaphragm Pump
Leverandør: VACUUBRAND
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
25743750
VACU25743750EA 114050 DKK
VACU25743750
VARIO® MD 12C NT VARIO select Chemistry Diaphragm Pump
Pumper Membranpumper, vakuum
Chemistry diaphragm pumps from VACUUBRAND for aggressive gases and vapors are available in the basic version, with two-point or VARIO® control. Their construction with fluoropolymers makes them very resistant to chemical vapors from inlet to exhaust.
  • VACUU·SELECT vacuum controller with graphical user interface and predefined applications simplifies work in the laboratory
  • Automatic boiling point detection and adjustment of the vacuum for short process times
  • Meets high vacuum requirements for large quantities of most high boiling solvents
  • Extraordinary