Safety spectacles, i-works
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9194.171 9194.175 9194.270 9194.365
UVEX9194.171EA 165 DKK
UVEX9194.171 UVEX9194.175 UVEX9194.270 UVEX9194.365
Safety spectacles, i-works
Briller
The 100% metal free uvex i-works encapsulates the best of uvex eyewear: the sporty design, panoramic lens and low weight make it extremely comfortable and together with the unrivalled permanent uvex lens coating technology (uvex supravision excellence), provides the highest level of optical clarity remaining fog free even after repeated cleaning. High-tech soft grip side arm materials further enhance comfort levels delivering a stable, non-slip fit.
  • Sporty design and panoramic lens for great lateral vision
  • High performance anti-fog coating on the inside,