Anti-LY-6H Mouse Monoclonal Antibody [clone: 9G379]

Leverandør: US Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

L7715-38A
USBIL7715-38AEA 4490 DKK
USBIL7715-38A
Anti-LY-6H Mouse Monoclonal Antibody [clone: 9G379]
Antistoffer
Recommended dilution: Immunochemistry: 2?g/ml

Type: Primary
Antigen: LY-6H
Clonality: monoclonal
Clone: 9G379
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR