Anti-LEP Rabbit Polyclonal Antibody

Leverandør: US Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

L1670-21A
USBIL1670-21AEA 4330 DKK
USBIL1670-21A
Anti-LEP Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: LEP (leptin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Bestil nu

Specifikationer

Antigen symbol LEP
Environmentally Preferable
Antigen name leptin
Antistof type Primary
Clonality Polyclonal
Konjugation Unconjugated
Reactivity Rat
Vært Rabbit
Isotype
Western blot Yes
ELISA Yes
Form lyophilized
Purification purified by immunoaffinity chromatography
Koncentration 1 mg/ml
Storage buffer 1mg/ml 0,05M phosphate buffer, 0,1M sodiumchloride, pH 7,2.
Opbevaringstemperatur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR