Dialysis tubular membrane, Cellu•Sep®
Leverandør: Uptima
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
T1-19-15
UPTIT1-19-15EA 0 DKK
UPTIT1-19-15
Dialysis tubular membrane, Cellu•Sep®
Slanger Dialyseslanger
Cellu·Sep® regenerated cellulose tubular membranes have proven time after time to be the smartest value in dialysis tubing available to laboratories anywhere around the globe.
Leveringsinformation: Each box of membrane is supplied with technical manual with instructions, analytical data sheets, disposable scissors and locking dialysis membrane clamps.
Emballering: Each box shipped in a sealed zip bag with the desiccant to control excess humidity.