3-Brom-5-nitrobenzoesyre

Leverandør: Uptima
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

QK628
UPTIQK628EA 0 DKK
UPTIQK628
3-Brom-5-nitrobenzoesyre
3-Brom-5-nitrobenzoesyre
Formel: C₇H₄BrNO₄
Molekylvægt: 246.02 g/mol
MDL Number: MFCD00100098
CAS-nummer: 6307-83-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR