Enclosed Waste Bins
Leverandør: TEKNOMEK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HWWB-FSFE HWWB-FSFEFOL HWWB-FSFEFOLC HWWB-FSFEL HWWB-MBFEFOL
TEKMHWWB-FSFEEA 0 DKK
TEKMHWWB-FSFE TEKMHWWB-FSFEFOL TEKMHWWB-FSFEFOLC TEKMHWWB-FSFEL TEKMHWWB-MBFEFOL
Enclosed Waste Bins
Beholdere Affaldsbeholdere
The enclosed waste bins lid is supplied with tough elastic bands.
  • Brushed dull polish finish
  • Mobile version available
  • Strong and sturdy design
  • Supplied ready to use
  • Foot operated lids available for easy operation
Our enclosed waste bins are manufactured from 304-grade stainless steel. A plastic part free design and food grade edge trims ensure our products are easy to clean, supporting rigorous hygiene standards. A tough elastic band securely holds the bag in place, reducing the risk of tearing or spilling waste.
Mobile versions that offer evenMore Product Information