Tetrabutylammonium acetate ≥90.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Tetra-n-butylammonium acetate

T2694-25G T2694-100G
TCIAT2694-25GEA 854 DKK
TCIAT2694-25G TCIAT2694-100G
Tetrabutylammonium acetate ≥90.0% (by titrimetric analysis)
Tetrabutylammonium acetate
Formel: C₁₈H₃₉NO₂
Smeltepunkt: 98°C
MDL Number: MFCD00043208
CAS-nummer: 10534-59-5
EINECS: 234-101-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=405882971c2595b7b46a8ce7bf9cbe65b34f36dc&vid=405882971c2595b7b46a8ce7bf9cbe65b34f36dc&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow powder to lump
Purity (Nonaqueous Titration) min. 90.0 %
Water max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR